Learn More
Arbor Assays Donkey Anti-Sheep IgG (H&L), 10MG
SDP
Catalog No. 502278565
Encompass_Preferred
An affinity purified Donkey Antibody to Sheep IgG,(H&L). Supplied in PBS with azide as preservative. Affinity purified on a column with covalently coupled Sheep IgG attached.
This item is not returnable. View return policy
Catalog No. 50-227-8565 Supplier Arbor Assays Supplier No. A01010MG
An affinity purified Donkey Antibody to Sheep IgG,(H&L). Supplied in PBS with azide as preservative. Affinity purified on a column with covalently coupled Sheep IgG attached.