Learn More
Immunology Consultants anti-Sheep IgG h+l Antibody (Donkey) - Affinity Purified
SDP
Catalog No. 502248887
Encompass_Preferred
This affinity‑purified donkey antibody recognizes both heavy and light chains of sheep IgG, providing broad, sensitive detection across a wide range of assay formats. Its strong specificity and low nonspecific binding support reliable performance in ELISA, Western blotting, immunohistochemistry, and other secondary antibody applications. The donkey host species reduces background associated with bovine or goat‑derived reagents, improving clarity in complex samples. DGHL‑35A’s consistent purification and robust binding characteristics make it a versatile secondary antibody for research, diagnostic development, and workflows requiring comprehensive detection of sheep IgG.
This item is not returnable. View return policy
Catalog No. 50-224-8887 Supplier Immunology Consultants Supplier No. DGHL35A
This affinity‑purified donkey antibody recognizes both heavy and light chains of sheep IgG, providing broad, sensitive detection across a wide range of assay formats. Its strong specificity and low nonspecific binding support reliable performance in ELISA, Western blotting, immunohistochemistry, and other secondary antibody applications. The donkey host species reduces background associated with bovine or goat‑derived reagents, improving clarity in complex samples. DGHL‑35A’s consistent purification and robust binding characteristics make it a versatile secondary antibody for research, diagnostic development, and workflows requiring comprehensive detection of sheep IgG.