Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Donkey aRb IgG Anti Secondary

Catalog No. 5000087071
Encompass_Preferred

Product Name: Unconjugated Donkey Anti-Rabbit IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Unconjugated Donkey Anti-Rabbit IgG (H&L)
Cytek Catalog Number: 70-8037-U500
Marker: IgG (H&L)
Target Species: Rabbit
Reported Cross Reactivity: Rabbit
Clonality: Polyclonal
Host Species: Donkey
Conjugation: Unconjugated
Size: 500 μg

Catalog No. 50-000-87071 Supplier Cytek Biosciences Inc Supplier No. 708037U500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: Unconjugated Donkey Anti-Rabbit IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Unconjugated Donkey Anti-Rabbit IgG (H&L)
Cytek Catalog Number: 70-8037-U500
Marker: IgG (H&L)
Target Species: Rabbit
Reported Cross Reactivity: Rabbit
Clonality: Polyclonal
Host Species: Donkey
Conjugation: Unconjugated
Size: 500 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.