Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Donkey aSheep IgG Anti

Catalog No. 5000087339
Encompass_Preferred

Product Name: HRP Donkey Anti-Sheep IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Donkey Anti-Sheep IgG (H&L)
Cytek Catalog Number: 72-8094-M001
Marker: IgG (H&L)
Target Species: Sheep
Reported Cross Reactivity: Sheep
Clonality: Polyclonal
Host Species: Donkey
Conjugation: HRP
Size: 1 mg

Catalog No. 50-000-87339 Supplier Cytek Biosciences Inc Supplier No. 728094M001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: HRP Donkey Anti-Sheep IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Donkey Anti-Sheep IgG (H&L)
Cytek Catalog Number: 72-8094-M001
Marker: IgG (H&L)
Target Species: Sheep
Reported Cross Reactivity: Sheep
Clonality: Polyclonal
Host Species: Donkey
Conjugation: HRP
Size: 1 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.