Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure F(ab')₂ Fragment Donkey Anti-Mouse IgG (H+L), .3ml
SDP
Supplier:  Jackson Immuno Research Labs 715036151
Encompass_Preferred
Peroxidase-affiniPure F(ab')2 Fragment Donkey anti-Mouse IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Rat,Shp Sr Prot) Purity: The antibody was purified from antisera by a combination of pepsin digestion and immunoaffinity chromatography using antigens coupled to agarose beads. Fc fragments and whole IgG molecules have been removed. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum albumin (IgG-Free, Protease-Free) Unit: 0.3 ml
Catalog No. NC9827055