Learn More
Rockland Immunochemicals Donkey Serum Non-Sterile 50 mL
SDP
Catalog No. 501943967
Encompass_Preferred
Donkey serum is used as a component of bioassays immunoassays or enzyme assays. Donkey serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Donkey serum is also available as a sterile preparation for use as a cell culture supplement.
Catalog No. 50-194-3967 Supplier Rockland Immunochemicals Supplier No. D220000050
Donkey serum is used as a component of bioassays immunoassays or enzyme assays. Donkey serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Donkey serum is also available as a sterile preparation for use as a cell culture supplement.