Learn More
Rockland Immunochemicals DONKEY SERUM (sterile) - D120-00-0050
SDP
Catalog No. 501058906
Encompass_Preferred
Sterile Donkey Serum is used as a supplement to cell culture media. Donkey Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
Catalog No. 50-105-8906 Supplier Rockland Immunochemicals Supplier No. D120000050
Sterile Donkey Serum is used as a supplement to cell culture media. Donkey Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
Product Title
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.