Learn More
Alkali Scientific ASI™ Donor Donkey Serum, 500mL
SDP
Catalog No. 503029133
Encompass_Preferred
Donor donkey serum is collected aseptically, from normal healthy donor animals. Donkey serum is commonly used as a blocking buffer for immunoassays such as immunohistochemistry, immunoblotting, and immunofluorescence.  Sterile-filtered Ships on ice. Store frozen at -20 to -10°C
Catalog No. 50-302-9133 Supplier Alkali Scientific Supplier No. FB33
Donor donkey serum is collected aseptically, from normal healthy donor animals. Donkey serum is commonly used as a blocking buffer for immunoassays such as immunohistochemistry, immunoblotting, and immunofluorescence.  Sterile-filtered Ships on ice. Store frozen at -20 to -10°C