Learn More
Bioworld Donor Equine serum, sterile filtered, 0.2 absolute, 100 mL
Catalog No. 50254220
Encompass_Preferred
Sterility tested.Mycoplasma testing upon request.Endotoxin level: < 0.5 ng/ml.Sterile filtered to 0.2 µmStorage Conditions: -20°C + 5°C.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-254-220 Supplier Bioworld Supplier No. 306111192
Sterility tested.Mycoplasma testing upon request.Endotoxin level: < 0.5 ng/ml.Sterile filtered to 0.2 µmStorage Conditions: -20°C + 5°C.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.