Learn More
Bioworld Donor Goat Serum, 100 mL
Catalog No. 50254190
Encompass_Preferred
Donor caprine serum is especially intended for use as a diluent/buffer in ELISA test kits. It is ideal as a blocking agent for Western blots and is also useful as a tissue culture media supplement.SpecificationsSterile-filteredShips on dry ice.
Catalog No. 50-254-190 Supplier Bioworld Supplier No. 306110821
Donor caprine serum is especially intended for use as a diluent/buffer in ELISA test kits. It is ideal as a blocking agent for Western blots and is also useful as a tissue culture media supplement.SpecificationsSterile-filteredShips on dry ice.