Learn More
Bioworld Donor Goat serum, sterile filtered, 0.2 absolute, heat inactivated, 100 mL
Catalog No. 50254221
Encompass_Preferred
Specially formulated for use as a diluent/buffer in ELISA test kits.Ideal as a blocking agent in Western blotting.Useful as tissue culture media supplement.Sterility tested.Endotoxin level: < 0.5 EU/ml.Mycoplasma testing upon request.Sterile filtered to 0.2 µmStorage Conditions: -20°C + 5°C.
Catalog No. 50-254-221 Supplier Bioworld Supplier No. 306111211
Specially formulated for use as a diluent/buffer in ELISA test kits.Ideal as a blocking agent in Western blotting.Useful as tissue culture media supplement.Sterility tested.Endotoxin level: < 0.5 EU/ml.Mycoplasma testing upon request.Sterile filtered to 0.2 µmStorage Conditions: -20°C + 5°C.