Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Donor Horse Serum, United States Origin
GREENER_CHOICE

Catalog No. MT35030CI Shop All Corning Products
Change view
Click to view available options
Serum Treatment:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Serum Treatment
MT35030CI None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT35030CI Supplier Corning™ Supplier No. 35030CI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Collected from a closed, controlled standing herd where each donor horse tested negative for Equine Infectious Anemia (EIA) per the Coggins Test.

True pooled and 0.2 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Equine
Form Liquid
Serum Treatment None
Sterility Non-sterile
Source United States
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.