Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Donor Vial Caddy

Catalog No. 22505276
Change view
Click to view available options
Dimensions (L x W x H):
7 x 4.25 x 1.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22-505-276 7 x 4.25 x 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-276 Supplier Marketlab Supplier No. 13459
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keeps blood samples together and organized during handling.

  • Lightweight caddy securely holds tubes before and after sample collection
  • Makes scanning barcodes easy and more efficient
  • Swivel clip allows attachment to blood bag
  • Designed for be reusable for cost-savings
  • Holds up to six 13mm or 16 mm tubes
  • Easy to clean with bleach solution

Specifications

Dimensions (L x W x H) 7 x 4.25 x 1.5 in.
Material Acrylic
Product Type Phlebotomy Tray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.