Learn More
Lonza Walkersville DonorPlex Heps 20 Donor mixed
Catalog No. 5000496313
Encompass
A consistent and robust source of pooled-donor human hepatocytes is important for DMPK studies With our new proprietary algorithm we are able to predict yield viability and functional phenotype of DonorPlex Human Hepatocytes which in turn will support more consistent results in your DMPK Each batch of pooled 20-donor mixed-gender human hepatocytes is characterized for viable yield and CYP1A2 CYP2A6 CYP2B6 CYP2C9 CYP2C19 CYP2E1 CYP3A4 UGT SULT OAT1B1/3 OCT and NTCP activities With our large inventory of single-donor human hepatocyte lots and using our proprietary algorithm to predict phenotype we can also custom-produce a pooled-donor hepatocyte batch with the phenotype to fit your needs The DonorPlex Hepatocytes - suspension-qualified hepatocytes are ideally suited for short-term ( 4hrs) in vitro drug clearance assays The suspension-qualified hepatocytes are not suitable for applications that require hepatocytes cultured in adhered monolayer
Catalog No. 50-004-96313 Supplier Lonza Walkersville Supplier No. HUCS20P
A consistent and robust source of pooled-donor human hepatocytes is important for DMPK studies With our new proprietary algorithm we are able to predict yield viability and functional phenotype of DonorPlex Human Hepatocytes which in turn will support more consistent results in your DMPK Each batch of pooled 20-donor mixed-gender human hepatocytes is characterized for viable yield and CYP1A2 CYP2A6 CYP2B6 CYP2C9 CYP2C19 CYP2E1 CYP3A4 UGT SULT OAT1B1/3 OCT and NTCP activities With our large inventory of single-donor human hepatocyte lots and using our proprietary algorithm to predict phenotype we can also custom-produce a pooled-donor hepatocyte batch with the phenotype to fit your needs The DonorPlex Hepatocytes - suspension-qualified hepatocytes are ideally suited for short-term ( 4hrs) in vitro drug clearance assays The suspension-qualified hepatocytes are not suitable for applications that require hepatocytes cultured in adhered monolayer