Learn More
Lonza Walkersville DonorPlex Heps 50 Donor Mixed
Catalog No. 5000496308
Encompass
A consistent and robust source of pooled donor human hepatocytes is important for DMPK studies With our new proprietary algorithm we are able to predict yield viability and functional phenotype representing the average human populations The DonorPlex Pooled Donor Human Hepatocytes 50-donor mixed gender is a roughly 50 50 (max 70 30) mixed gender 50-donor pool designed to function like the average human liver Each batch is characterized for viable yield and CYP1A2 CYP2A6 CYP2B6 CYP2C9 CYP2C19 CYP2E1 CYP3A4 UGT SULT OAT1B1/3 OCT and NTCP activities With our large inventory of single donor human hepatocyte lots and using our proprietary algorithm to predict phenotype we can also custom-produce a pooled donor hepatocyte batch with the phenotype to fit your needs
Catalog No. 50-004-96308 Supplier Lonza Walkersville Supplier No. HUCS50P
A consistent and robust source of pooled donor human hepatocytes is important for DMPK studies With our new proprietary algorithm we are able to predict yield viability and functional phenotype representing the average human populations The DonorPlex Pooled Donor Human Hepatocytes 50-donor mixed gender is a roughly 50 50 (max 70 30) mixed gender 50-donor pool designed to function like the average human liver Each batch is characterized for viable yield and CYP1A2 CYP2A6 CYP2B6 CYP2C9 CYP2C19 CYP2E1 CYP3A4 UGT SULT OAT1B1/3 OCT and NTCP activities With our large inventory of single donor human hepatocyte lots and using our proprietary algorithm to predict phenotype we can also custom-produce a pooled donor hepatocyte batch with the phenotype to fit your needs