Learn More
Aviva Systems Biology Corp DONSON Antibody - middle region (ARP45862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DONSON. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DONSON. Alternate names: B17, C21orf60, DKFZP434M035, C2TAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53457