Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl Donut Bumper

Catalog No. 11222177
Change view
Click to view available options
Diameter (English):
3.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
11-222-177 3.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-222-177 Supplier Wanzl Supplier No. T.DRB325
Add to Cart
Add to Cart
This item is not returnable. View return policy

Donut Bumper fits over caster, on post to protect walls and other units from damage.

  • Add-on option for additional customization
  • Fits over caster to protect walls and other units from damage

Specifications

For Use With (Application) Protect Walls and Other Units From Damage
Includes 1 Bumper
For Use With (Equipment) Casters
With Casters No
Color Chrome
Diameter (English) 3.25 in.
Material Rubber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.