Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Doodlebug™ Easy Erasing Pad

Catalog No. 17623891 Shop All 3M Products
Change view
Click to view available options
Color:
Blue, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-891 Blue, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-891 Supplier 3M™ Supplier No. 7000126851
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Doodlebug™ Easy Erasing Pad is ideal for removing stains, marks, ink and more. Just use water; no chemicals or sprays needed. Fits 3M™ Doodlebug™ Handblock Pad Holder 6473 or 3M™ Doodlebug™ Pad Holder 6472.

  • Pad will not scratch glass, ceramic tile, fiberglass, porcelain, Formica™ Laminate, metal, DuPont™ Corian™ Solid Surface, marble, stainless steel, plastic, vinyl, and most wall coatings
  • Soft, flexible - use with water. Detergents and sanitizing cleaners not recommended for use with pad
  • Pad will not scratch glass, ceramic tile, fiberglass, porcelain, Formica™ Laminate metal, DuPont™ Corian™ Sold Surface
  • Use on white boards, sports equipment, countertops, flat painted surfaces, tables, chairs, walls, stainless steel, hard and textured surfaces, vinyl seating, sneeze guards.
  • Detergents and sanitizing cleaners are not recommended for use with this product
TRUSTED_SUSTAINABILITY

Specifications

Product Type Scrub Pad
Color Blue, White
For Use With (Application) Remove Stains, Marks, Ink and More
Length (English) 10 in.
Length (Metric) 254 mm
Model 4610
Quantity 20/Cs.
Width (English) 4.625 in.
Width (Metric) 117 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.