Learn More
Medchemexpress LLC Dooku1 | 2253744-54-4 | 99.9% | 326.20 g/mol | C13H9Cl2N3OS | 10MG
SDP
Catalog No. 5000237853
Encompass_Preferred
Dooku1 is a small-molecule antagonist of Yoda1-activated Piezo1 mechanosensitive ion channels used in vascular physiology and mechanotransduction research. It antagonizes Yoda1-induced Piezo1 activation and inhibits Yoda1-evoked aortic relaxation. Supplied as a solid or as a DMSO solution, it is suitable for biochemical and cellular assays; chemical formula C13H9Cl2N3OS, molecular weight 326.20 g/mol, stated purity 99.93%.
  • Selective antagonist of Yoda1-activated Piezo1 channels.
  • Demonstrated activity in cell-based assays and aortic tissue studies.
  • Supplied as solid powder or 10 mM solution in DMSO for convenience.
  • High stated purity suitable for research applications.
  • Available in multiple packaging sizes for scale-up studies.
Catalog No. 50-002-37853 Supplier Medchemexpress LLC Supplier No. HY12601010MG
Dooku1 is a small-molecule antagonist of Yoda1-activated Piezo1 mechanosensitive ion channels used in vascular physiology and mechanotransduction research. It antagonizes Yoda1-induced Piezo1 activation and inhibits Yoda1-evoked aortic relaxation. Supplied as a solid or as a DMSO solution, it is suitable for biochemical and cellular assays; chemical formula C13H9Cl2N3OS, molecular weight 326.20 g/mol, stated purity 99.93%.
  • Selective antagonist of Yoda1-activated Piezo1 channels.
  • Demonstrated activity in cell-based assays and aortic tissue studies.
  • Supplied as solid powder or 10 mM solution in DMSO for convenience.
  • High stated purity suitable for research applications.
  • Available in multiple packaging sizes for scale-up studies.