Learn More
Medchemexpress LLC Dooku1 | 2253744-54-4 | 99.9% | 326.20 g/mol | C13H9Cl2N3OS | 25MG
SDP
Supplier:  Medchemexpress LLC HY12601025MG
Encompass_Preferred
Dooku1 is a reversible small-molecule antagonist of Yoda1-activated Piezo1 channels used to study Piezo1-mediated Ca2+ entry and vascular physiology. It has documented cellular activity in endothelial and HEK cell assays and is supplied as a high-purity research compound with specified solubility and storage recommendations.
Catalog No. 50-002-37851