Learn More
Selleck Chemical LLC Dooku1-5mg
SDP
Supplier:  Selleck Chemical LLC S29535MG
Encompass_Preferred
Dooku1 is an analog of Yoda1 with antagonist activity against mechanosensitive Piezo1 channel. Dooku1 inhibits 2 μM Yoda1-induced Ca2+-entry with IC50 of 1.3 μM in HEK 293 cells and 1.5 μM in HUVECs, respectively.
Catalog No. 50-238-9720