Learn More
Medchemexpress LLC HY-126010 5mg Medchemexpress, Dooku1 CAS:2253744-54-4 Purity:>98%
SDP
Catalog No. 501969714
Encompass_Preferred
Medchemexpress, HY-126010 5mg Dooku1 CAS:2253744-54-4 Dooku1, an analog of Yoda1, is a selective antagonist of the endogenous Piezo1 channel. Dooku1 inhibited 2 μM Yoda1-induced Ca2+-entry with IC50 values of 1.3 μM (in HEK 293 cells) and 1.5 μM (in HUVECs). Dooku1 inhibits Yoda1-induced relaxation of aorta. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9714 Supplier Medchemexpress LLC Supplier No. HY1260105MG
Medchemexpress, HY-126010 5mg Dooku1 CAS:2253744-54-4 Dooku1, an analog of Yoda1, is a selective antagonist of the endogenous Piezo1 channel. Dooku1 inhibited 2 μM Yoda1-induced Ca2+-entry with IC50 values of 1.3 μM (in HEK 293 cells) and 1.5 μM (in HUVECs). Dooku1 inhibits Yoda1-induced relaxation of aorta. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.