Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Door Gasket
SDP

Catalog No. 08111646 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers FDL 115
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-111-646 BINDER Chambers FDL 115
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-111-646 Supplier BINDER™ Supplier No. 80120499

missing translation for 'validMainframeMsg'

Material: Silicone; For Use With: BINDER units FDL 115

  • Manufactured from silicone
  • Gasket wears out more quickly in temperatures above 250°C
  • For use in FDL 115 Safety Drying Chambers

Specifications

Description Door gasket - made from silicone, resistant to high temperatures for operating temperatures above 200°C. Note: The gasket wears out more quickly in temperatures above 250°C
For Use With (Equipment) BINDER Chambers FDL 115
Material Silicone
Temperature Range (Metric) Ambient to 250°C
Type Door Gasket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.