Learn More
BINDER™ Door Gasket FKM
SDP
Catalog No. 08111956 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Model VD/VDL 115
Catalog No. For Use With (Equipment)
08-111-956 Model VD/VDL 115
1 options
Catalog No. 08-111-956 Supplier BINDER™ Supplier No. 86120009
Add to Cart
Add to Cart
Door Gasket resists temperatures up to 150°C.
  • Option for BINDER series VD/VDL
  • Door gasket made of FKM (a fluoroelastomer)
  • Silicone free
  • Resistant to high temperatures and many chemical substances and is used in cases were silicone fails to meet the specifications.
Specifications
Chamber Material FKM
For Use With (Equipment) Model VD/VDL 115
Material FKM
Type Door Gasket