Learn More
MSC 80/20 Inc. Door Slide Track, 10 Series, Aluminum, 1219.2 mm, 1 ea.
Catalog No. 5000512875
Encompass
This lower door slide track is designed for 10 series profiles, providing a smooth glide for panels. The track snaps directly into a T-slot, requiring no additional hardware for assembly, and is typically paired with an upper door slide track to allow for easy panel insertion after frame construction.
  • Features a radius bearing surface for smooth panel movement
  • Constructed from durable, clear anodized aluminum
  • Snaps into T-slots with no required hardware
  • Supports a door thickness of 0.281 in.
  • Measures 48 in. (1,219.2 mm) in length
Catalog No. 50-005-12875 Supplier MSC Supplier No. 76824408
This lower door slide track is designed for 10 series profiles, providing a smooth glide for panels. The track snaps directly into a T-slot, requiring no additional hardware for assembly, and is typically paired with an upper door slide track to allow for easy panel insertion after frame construction.
  • Features a radius bearing surface for smooth panel movement
  • Constructed from durable, clear anodized aluminum
  • Snaps into T-slots with no required hardware
  • Supports a door thickness of 0.281 in.
  • Measures 48 in. (1,219.2 mm) in length