Learn More
Cayman Chemical Dopamine-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 203633-19-6, 10 mg
SDP
Catalog No. 502912945
Encompass_Preferred
An internal standard for the quantification of dopamine by GC- or LC-MS
Catalog No. 50-291-2945 Supplier Cayman Chemical Supplier No. 3630910 MG
An internal standard for the quantification of dopamine by GC- or LC-MS