Learn More
Medchemexpress LLC Dopamine-13C,15N hydrochloride | 369656-74-6 | 99.0% | 1 MG
SDP
Catalog No. 5000408248
Encompass_Preferred
Dopamine-13C,15N hydrochloride is a stable isotope-labeled form of Dopamine hydrochloride, incorporating 13C and 15N isotopes. This compound is intended for research use only and not for human application. It is primarily utilized in scientific studies, particularly where precise tracing and quantitative analysis are required.
  • Labeled with 13C and 15N isotopes.
  • Used as a tracer in research.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in techniques like NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08248 Supplier Medchemexpress LLC Supplier No. HYB0451AS61MG
Dopamine-13C,15N hydrochloride is a stable isotope-labeled form of Dopamine hydrochloride, incorporating 13C and 15N isotopes. This compound is intended for research use only and not for human application. It is primarily utilized in scientific studies, particularly where precise tracing and quantitative analysis are required.
  • Labeled with 13C and 15N isotopes.
  • Used as a tracer in research.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in techniques like NMR, GC-MS, or LC-MS.