Learn More
Medchemexpress LLC Dopamine-13C,15N hydrochloride | 369656-74-6 | 99.0% | 5 MG
SDP
Catalog No. 5000405951
Encompass_Preferred
Dopamine-13C,15N (hydrochloride) is the 13C and 15N labeled Dopamine hydrochloride. It is intended for research use only and not for sale to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has also garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Dopamine-13C,15N (hydrochloride) is the 13C and 15N labeled Dopamine hydrochloride.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05951 Supplier Medchemexpress LLC Supplier No. HYB0451AS65MG
Dopamine-13C,15N (hydrochloride) is the 13C and 15N labeled Dopamine hydrochloride. It is intended for research use only and not for sale to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has also garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Dopamine-13C,15N (hydrochloride) is the 13C and 15N labeled Dopamine hydrochloride.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.