Learn More
Medchemexpress LLC Dopamine-13C hydrochloride | 335081-04-4 | 99.7% | 5 MG
SDP
Catalog No. 5000405950
Encompass_Preferred
Dopamine-13C (hydrochloride) is the 13C labeled Dopamine hydrochloride (ASL279). It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05950 Supplier Medchemexpress LLC Supplier No. HYB0451AS55MG
Dopamine-13C (hydrochloride) is the 13C labeled Dopamine hydrochloride (ASL279). It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS