Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dopamine 4-sulfate | 38339-02-5 | 96.0% | 233.24 g/mol | C8H11NO5S | 5 MG
SDP

Catalog No. 5000263768
Encompass_Preferred

Dopamine 4-sulfate is a sulfated metabolite of the neurotransmitter dopamine used as a research reagent and metabolite reference for studies of dopamine metabolism and sulfotransferase activity. It is supplied as a high-purity solid for biochemical, analytical, and in vitro applications.

  • Used as a metabolite reference in biochemical and pharmacological assays.
  • Chemical formula C8H11NO5S; molecular weight 233.24 g/mol.
  • Purity approximately 96% as supplied.
  • Available in small research sizes: 1 mg, 5 mg, 10 mg, 50 mg.
  • Suitable for analytical standards and in vitro studies.

Catalog No. 50-002-63768 Supplier Medchemexpress LLC Supplier No. HY1132575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dopamine 4-sulfate is a sulfated metabolite of the neurotransmitter dopamine used as a research reagent and metabolite reference for studies of dopamine metabolism and sulfotransferase activity. It is supplied as a high-purity solid for biochemical, analytical, and in vitro applications.

  • Used as a metabolite reference in biochemical and pharmacological assays.
  • Chemical formula C8H11NO5S; molecular weight 233.24 g/mol.
  • Purity approximately 96% as supplied.
  • Available in small research sizes: 1 mg, 5 mg, 10 mg, 50 mg.
  • Suitable for analytical standards and in vitro studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.