Learn More
Medchemexpress LLC Dopamine D2 receptor agonist-3 | 1257326-24-1 | 98.3% | 295.81 | C15H22ClN3O | 10 MG
SDP
Catalog No. 5000246916
Encompass_Preferred
Dopamine D2 receptor agonist-3 is a research compound described as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist. It is supplied as a solid powder with documented purity and analytical data for use in in vitro and preclinical receptor pharmacology research.
  • High purity of 98.3%.
  • Molecular formula C15H22ClN3O and molecular weight 295.81.
  • Selective D2 partial agonist and D3 antagonist (pEC50 = 8.3 and <5.5).
  • Supplied as a solid powder for research use only.
  • Available in multiple sizes, including 10 MG packaging.
  • Datasheet and certificate of analysis available.
Catalog No. 50-002-46916 Supplier Medchemexpress LLC Supplier No. HY10341810MG
Dopamine D2 receptor agonist-3 is a research compound described as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist. It is supplied as a solid powder with documented purity and analytical data for use in in vitro and preclinical receptor pharmacology research.
  • High purity of 98.3%.
  • Molecular formula C15H22ClN3O and molecular weight 295.81.
  • Selective D2 partial agonist and D3 antagonist (pEC50 = 8.3 and <5.5).
  • Supplied as a solid powder for research use only.
  • Available in multiple sizes, including 10 MG packaging.
  • Datasheet and certificate of analysis available.