Learn More
Medchemexpress LLC Dopamine D2 receptor agonist-3 | 1257326-24-1 | 98.3% | 295.81 | C15H22ClN3O | 5 MG
SDP
Catalog No. 5000246493
Encompass_Preferred
Dopamine D2 receptor agonist-3 is a small-molecule research compound that acts as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist, with reported in vitro potency and demonstrated brain penetration in vivo. It is supplied as a solid (white to off-white) with high stated purity for laboratory use.
  • Selective D2 partial agonist and D3 antagonist activity.
  • Potency reported as pEC50 D2 = 8.3; D3 < 5.5 (approx. D2 EC50 ≈ 5 nM).
  • Demonstrated brain penetration (brain:blood ratio 5.5; Tmax 0.5 h at 3 mg/kg, s.c.).
  • High purity (98.3%) suitable for research applications.
  • Solid, white to off-white appearance, available in small research pack sizes.
Catalog No. 50-002-46493 Supplier Medchemexpress LLC Supplier No. HY1034185MG
Dopamine D2 receptor agonist-3 is a small-molecule research compound that acts as a selective dopamine D2 receptor partial agonist and a dopamine D3 receptor antagonist, with reported in vitro potency and demonstrated brain penetration in vivo. It is supplied as a solid (white to off-white) with high stated purity for laboratory use.
  • Selective D2 partial agonist and D3 antagonist activity.
  • Potency reported as pEC50 D2 = 8.3; D3 < 5.5 (approx. D2 EC50 ≈ 5 nM).
  • Demonstrated brain penetration (brain:blood ratio 5.5; Tmax 0.5 h at 3 mg/kg, s.c.).
  • High purity (98.3%) suitable for research applications.
  • Solid, white to off-white appearance, available in small research pack sizes.