Learn More
TARGETMOL CHEMICALS INC DOPAMINE D2 RECEPTOR ANT 5MG
SDP
Catalog No. 502581580
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dopamine D2 receptor antagonist-1 is a negative allosteric modulator (NAM) of dopamine D2 receptor (D2R) with sub-mM affinity. purity: 99%
Catalog No. 50-258-1580 Supplier TARGETMOL CHEMICALS INC Supplier No. T110775MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dopamine D2 receptor antagonist-1 is a negative allosteric modulator (NAM) of dopamine D2 receptor (D2R) with sub-mM affinity. purity: 99%