Learn More
Medchemexpress LLC Dopamine-d3 hydrochloride | 53587-30-7 | 99.9% | 1 MG
SDP
Catalog No. 5000405945
Encompass_Preferred
Dopamine-d3 (hydrochloride) is the deuterium-labeled form of Dopamine hydrochloride (ASL279). It is used for research purposes and is not sold to patients. This compound is described as a stable heavy isotope that can be incorporated into drug molecules, primarily as a tracer for quantitation during drug development. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
Catalog No. 50-004-05945 Supplier Medchemexpress LLC Supplier No. HYB0451AS41MG
Dopamine-d3 (hydrochloride) is the deuterium-labeled form of Dopamine hydrochloride (ASL279). It is used for research purposes and is not sold to patients. This compound is described as a stable heavy isotope that can be incorporated into drug molecules, primarily as a tracer for quantitation during drug development. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.