Learn More
Medchemexpress LLC Dopamine-d3 (hydrochloride) | 53587-30-7 | 99.9% | 5 MG
SDP
Catalog No. 5000406786
Encompass_Preferred
Dopamine-d3 (hydrochloride) is the deuterium labeled Dopamine hydrochloride (ASL279)-. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06786 Supplier Medchemexpress LLC Supplier No. HYB0451AS45MG
Dopamine-d3 (hydrochloride) is the deuterium labeled Dopamine hydrochloride (ASL279)-. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS