Learn More
Apexbio Technology LLC DOPAMINE-D4-HYDROCHLORIDE-10MG
SDP
Catalog No. 5000570309
Encompass_Preferred
Dopamine-d4 hydrochloride (CAS No 203633-19-6) is a deuterium-labeled derivative of dopamine commonly used in biomedical research to study dopaminergic pathways and neurotransmitter dynamics Originating from synthetic processes it acts primarily on dopamine receptors influencing pathways associated with neurological and psychiatric functions Its activity is typically observed in the nanomolar to low micromolar range making it suitable for precise mechanistic studies This compound is often utilized in vitro in cell models to investigate receptor interactions and in animal studies to explore behavioral and physiological effects It is also employed in drug screening programs to evaluate potential therapeutic agents targeting neurological conditions Experimental concentrations typically depend on the specific research design ensuring flexibility in various study protocols
Catalog No. 50-005-70309 Supplier Apexbio Technology LLC Supplier No. SL101710
Dopamine-d4 hydrochloride (CAS No 203633-19-6) is a deuterium-labeled derivative of dopamine commonly used in biomedical research to study dopaminergic pathways and neurotransmitter dynamics Originating from synthetic processes it acts primarily on dopamine receptors influencing pathways associated with neurological and psychiatric functions Its activity is typically observed in the nanomolar to low micromolar range making it suitable for precise mechanistic studies This compound is often utilized in vitro in cell models to investigate receptor interactions and in animal studies to explore behavioral and physiological effects It is also employed in drug screening programs to evaluate potential therapeutic agents targeting neurological conditions Experimental concentrations typically depend on the specific research design ensuring flexibility in various study protocols