Learn More
Abcam Dopamine ELISA Kit, 96TEST
Supplier:  Abcam AB28523896TEST
Encompass_Preferred
Dopamine ELISA kit is a competitive ELISA designed to quantify dopamine (DA) with a sensitivity of <0.938 ng/mL. - Specifically binds with DA - no apparent cross-reactivity with other analogues - Wide dynamic range – quantifies 1.56–100 ng/mL - Broad sample compatibility – works with serum, plasma, tissue homogenates, cell culture supernatants, and other biological fluids - Assay platform – microplate (12 x 8 well strips) for easy setup and reliable results
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5493