Learn More
Enzo Life Sciences Dopamine ELISA Kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1880001
Encompass_Preferred
Sensitive ELISA kit for the quantification of Dopamine in any species. The Dopamine ELISA kit is a colorimetric immunoassay kit with results in < 2 hours. Absorbance is read at 450 nm. Measure up to 40 samples in duplicate per 96 well  kit. Sensitivity: 0.938 ng/mL (1.56 - 100 ng/mL). Assay Time: < 2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Lyophilized Standard, Sample/standard dilution buffer, Concentrated biotin detector antibody, Antibody Dilution Buffer, HRP-Streptavidin Conjugate, Conjugate Dilution Buffer, TMB Substrate, Stop Solution, Wash Buffer (25X), Plate Sealer
Catalog No. NC1597282