Learn More
Apexbio Technology LLC Dopamine HCl 62-31-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14825.1
Encompass_Preferred
Dopamine HCl (CAS 62-31-7) is a catecholamine neurotransmitter that functions as an endogenous agonist at dopamine receptors (subtypes D1 D5) Mechanistically dopamine activates these G protein-coupled receptors modulating neurotransmission and influencing diverse physiological processes such as motor control cognition motivation and neuroendocrine regulation In biomedical research dopamine HCl serves as a standard reagent for investigating dopaminergic signaling pathways receptor pharmacology and the molecular basis of neurological disorders
Catalog No. 50-000-61749