Learn More
Medchemexpress LLC Dopamine hydrochloride | 62-31-7 | 99.79% | 1 ML
SDP
Catalog No. 5000406054
Encompass_Preferred
Dopamine hydrochloride (ASL279) is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays several important roles in the brain and body. Dopamine hydrochloride acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
Catalog No. 50-004-06054 Supplier Medchemexpress LLC Supplier No. HYB0451A10MM/1ML
Dopamine hydrochloride (ASL279) is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays several important roles in the brain and body. Dopamine hydrochloride acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.