Learn More
Medchemexpress LLC Dopamine hydrochloride | 62-31-7 | 99.8% | 100 MG
SDP
Catalog No. 5000407825
Encompass_Preferred
Dopamine hydrochloride (Standard) is an analytical standard of Dopamine (hydrochloride) intended for research and analytical applications. It is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain, playing important roles in the brain and body. It acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Serves as an analytical and reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Applicable with techniques such as HPLC, GC, and MS.
Catalog No. 50-004-07825 Supplier Medchemexpress LLC Supplier No. HYB0451AR100MG
Dopamine hydrochloride (Standard) is an analytical standard of Dopamine (hydrochloride) intended for research and analytical applications. It is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain, playing important roles in the brain and body. It acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Serves as an analytical and reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Applicable with techniques such as HPLC, GC, and MS.