Learn More
Medchemexpress LLC Dopamine hydrochloride | 62-31-7 | 99.8% | 50 MG
SDP
Catalog No. 5000406324
Encompass_Preferred
Dopamine hydrochloride (ASL279) is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays important roles in the brain and body by acting through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis. This product is intended for research use only.
  • Catecholamine neurotransmitter
  • Acts through D2 dopamine receptors
  • Induces endocytosis of VEGFR2
  • Critical for promoting angiogenesis
  • Inhibits VPF/VEGF-induced HUVEC proliferation
  • Selectively inhibits vascular permeabilizing and angiogenic activities of VPF/VEGF
Catalog No. 50-004-06324 Supplier Medchemexpress LLC Supplier No. HYB0451A50MG
Dopamine hydrochloride (ASL279) is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays important roles in the brain and body by acting through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis. This product is intended for research use only.
  • Catecholamine neurotransmitter
  • Acts through D2 dopamine receptors
  • Induces endocytosis of VEGFR2
  • Critical for promoting angiogenesis
  • Inhibits VPF/VEGF-induced HUVEC proliferation
  • Selectively inhibits vascular permeabilizing and angiogenic activities of VPF/VEGF