Learn More
Medchemexpress LLC Dopamine hydrochloride | 62-31-7 | 99.8% | 500 MG
Supplier: Medchemexpress LLC HYB0451A500MG
Dopamine hydrochloride is a catecholamine neurotransmitter that is produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays several important roles in the brain and body by acting through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
- Catecholamine neurotransmitter
- Acts through D2 dopamine receptors
- Induces endocytosis of VEGFR2
- Critical for promoting angiogenesis
- Inhibits VPF/VEGF-induced HUVEC proliferation
- Selectively inhibits vascular permeabilizing and angiogenic activities of VPF/VEGF