Learn More
Medchemexpress LLC Dopamine (hydrochloride) | 62-31-7 | MFCD00012898 | 99.8% | 100 MG
SDP
Catalog No. 5000405948
Encompass_Preferred
Dopamine hydrochloride is a catecholamine neurotransmitter produced in the brain's substantia nigra, ventral tegmental area, and hypothalamus. It plays important roles in the brain and body. This compound acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Catecholamine neurotransmitter
  • Produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain
  • Plays several important roles in the brain and body
  • Acts through D2 dopamine receptors
  • Induces endocytosis of VEGFR2
  • Critical for promoting angiogenesis
Catalog No. 50-004-05948 Supplier Medchemexpress LLC Supplier No. HYB0451A100MG
Dopamine hydrochloride is a catecholamine neurotransmitter produced in the brain's substantia nigra, ventral tegmental area, and hypothalamus. It plays important roles in the brain and body. This compound acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Catecholamine neurotransmitter
  • Produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain
  • Plays several important roles in the brain and body
  • Acts through D2 dopamine receptors
  • Induces endocytosis of VEGFR2
  • Critical for promoting angiogenesis