Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dopamine hydrochloride (standard) | 62-31-7 | 99.75% | 25 MG
SDP

Catalog No. 5000450992
Encompass_Preferred

Dopamine hydrochloride is an analytical standard intended for research and analytical applications. This catecholamine neurotransmitter is produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays important roles in the brain and body, acting through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis. The compound is used in qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analysis
  • Catecholamine neurotransmitter
  • Acts via D2 dopamine receptors
  • Promotes angiogenesis
  • Suitable for HPLC, GC, and MS experiments
  • Molecular weight: 189.64
  • Formula: C8H12ClNO2
  • Appearance: Solid, white to off-white
  • Classified as phenols and polyphenols
  • Derived from microorganisms
  • Endogenous metabolite

Catalog No. 50-004-50992 Supplier Medchemexpress LLC Supplier No. 25MGHYB0451AR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dopamine hydrochloride is an analytical standard intended for research and analytical applications. This catecholamine neurotransmitter is produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain. It plays important roles in the brain and body, acting through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis. The compound is used in qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analysis
  • Catecholamine neurotransmitter
  • Acts via D2 dopamine receptors
  • Promotes angiogenesis
  • Suitable for HPLC, GC, and MS experiments
  • Molecular weight: 189.64
  • Formula: C8H12ClNO2
  • Appearance: Solid, white to off-white
  • Classified as phenols and polyphenols
  • Derived from microorganisms
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.