Learn More
Medchemexpress LLC Dopamine hydrochloride (standard) | 62-31-7 | MFCD00012898 | 99.8% | 50 MG
SDP
Catalog No. 5000408242
Encompass_Preferred
Dopamine hydrochloride (standard) is an analytical standard intended for research and analytical applications. It is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain, playing several important roles in the brain and body. It acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Analytical standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-08242 Supplier Medchemexpress LLC Supplier No. HYB0451AR50MG
Dopamine hydrochloride (standard) is an analytical standard intended for research and analytical applications. It is a catecholamine neurotransmitter produced in the substantia nigra, ventral tegmental area, and hypothalamus of the brain, playing several important roles in the brain and body. It acts through D2 dopamine receptors to induce endocytosis of VEGFR2, which is critical for promoting angiogenesis.
  • Analytical standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications