Learn More
Enzo Life Sciences Dopamine receptor D1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905789100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat dopamine receptor D1. UniProt ID: P18901. Species reactivity: Rat. Creossreactivity: Predicted species reactivity based on epitope sequence homology: mouse, dog, bovine, wild boar. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-200-7056