Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dopamine Receptor D3 Antibody (YA1742) | 10 UL
SDP

Supplier:  Medchemexpress LLC HYP8199710UL

Encompass_Preferred

Dopamine Receptor D3 Antibody (YA1742) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Dopamine Receptor D3. Its function is mediated by G proteins which inhibit adenylyl cyclase and it promotes cell proliferation. It is supplied in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.

  • Host: Rabbit
  • Clonality: Recombinant, Monoclonal
  • Species reactivity: Human, Mouse, Rat
  • Immunogen: A synthesized peptide derived from human Dopamine Receptor D3 aa1-50/400
  • Application: WB (Western Blot) at 1:500-1:1000 dilution
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Unmodified
  • Isotype: IgG
  • Appearance: Liquid
  • Subcellular localization: Cell membrane; Multi-pass membrane protein
  • Expression tissue specificity: Brain

Catalog No. 50-004-41048


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.