Learn More
Medchemexpress LLC Dopamine Receptor D3 Antibody (YA1742) | 50 UL
SDP
Catalog No. 5000441658
Encompass_Preferred
Dopamine Receptor D3 Antibody (YA1742) is a rabbit-derived and non-conjugated IgG monoclonal antibody targeting Dopamine Receptor D3. It is reactive with human, mouse, and rat species, making it suitable for various research applications, particularly Western Blot (WB). The antibody is supplied as a liquid formulation containing rabbit IgG in 10mM phosphate-buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.
  • Targets dopamine receptor D3
  • Reactive with human, mouse, and rat species
  • Non-conjugated IgG monoclonal antibody
  • Application in Western blot (WB) with a dilution ratio of 1:500-1:1000
  • Predicted and observed band size of 44 kDa
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
Catalog No. 50-004-41658 Supplier Medchemexpress LLC Supplier No. HYP8199750UL
Dopamine Receptor D3 Antibody (YA1742) is a rabbit-derived and non-conjugated IgG monoclonal antibody targeting Dopamine Receptor D3. It is reactive with human, mouse, and rat species, making it suitable for various research applications, particularly Western Blot (WB). The antibody is supplied as a liquid formulation containing rabbit IgG in 10mM phosphate-buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.
  • Targets dopamine receptor D3
  • Reactive with human, mouse, and rat species
  • Non-conjugated IgG monoclonal antibody
  • Application in Western blot (WB) with a dilution ratio of 1:500-1:1000
  • Predicted and observed band size of 44 kDa
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles