Learn More
Alpco Ltd Dopamine Research ELISA
SDP
Supplier:  Alpco Ltd 17DOPHUE01RES
Encompass_Preferred
The Dopamine ELISA (Research) is an enzyme immunoassay intended for the quantitative determination of dopamine in Biological Fluids and Tissue Homogenate using sample volumes between 1-100 ul, depending on sample type and expected concentrations.
Catalog No. NC2299763